Learn More
Thermo Scientific™ Nalgene™ Dropper Bottles

Description
Choose Thermo Scientific™ Nalgene™ LDPE White Dropper Bottles for accurate, reliable, repeatable dispensing of various reagents and as an excellent alternative to pipetting devices. Nalgene™ Dropper Bottles are made of opaque LDPE and are ideal for UV lightsensitive solutions. Delivers 40μL drops (based on water; viscosity affects drop size), dispensed one drop at a time. Excellent chemical resistance; suitable for most biotech, diagnostic and pharmaceutical applications.
- Dropper control tip snaps into place for a secure fit
- Dispensing tip and closures included, choose from white or multi-colored caps
- Leakproof†
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity (Metric) | 15 mL |
Capacity (English) | 0.5 oz. |
Color | White |
Description | LDPE Dropper Bottle |
Includes | Dispensing tip and closures |
Material | LDPE |
No. per Case | 25 |
No. per Pack | 25 |
Closure Color | White |
Certifications/Compliance | ISO 13485:2003 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Documents
Product Information
Schematics & Diagrams
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7441058010 | Certificate of Compliance | 20/01/2025 | 2751-9050 |
7444968010 | Certificate of Compliance | 31/12/2024 | 2751-9050 |
7289765010 | Certificate of Compliance | 18/06/2018 | 2751-9050 |
7289136010 | Certificate of Compliance | 18/06/2018 | 2751-9050 |
7274978010 | Certificate of Compliance | 18/12/2017 | 2751-9050 |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.