missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes
GREENER_CHOICE

Use these friction-fit polypropylene closures featuring a convenient pull tab for easy removal from centrifuge tubes.

Brand:  Thermo Scientific™ DS3111-0029

 View more versions of this product

Product Code. 10564441

  • 48.10 € / Pack of 20
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Easily remove Thermo Scientific™ Nalgene™ Friction-Fit Polypropylene Closures for Plastic Centrifuge Tubes from the centrifuge with a convenient pull tab. These autoclavable closures are designed to fit Nalgene™ round and conical centrifuge tubes. They may not be liquid-tight during high-speed centrifugation.

Ordering Information:

DS3111-0030 fits non-lipped 50mL centrifuge tubes only.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

29 mm
Autoclavable
Polypropylene
20
Tube with 29 mm O.D.
Centrifuge Tube Cap
Designed to be compatible with Nalgene round and conical centrifuge tubes
20
Nalgene™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers