Learn More
Thermo Scientific™ CryoTube™ Holders
Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.
Brand: Thermo Scientific™ 376589
23.92 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.
- Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
- Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping
Specifications
Interlocking | |
Vial Holder | |
Nunc™ | |
4 x 10 |
PPO | |
40 Cryogenic Tubes | |
1/Pk., 15/Cs. |