Learn More
Thermo Scientific™ Nalgene™ Graduated HDPE Bucket

Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle.
Brand: Thermo Scientific™ 7002-0025
90.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle. This heavy-duty, rugged high density polyethylene bucket is chemically compatible with a wide range of lab reagents.
- High-density polyethylene construction offers more durability than hard rubber
- Polyethylene-coated wire bail handle provides a comfortable and sturdy grip
- Molded-in graduations in pints and liters are easy to read
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications
9.5 L | |
HDPE | |
2.5 gal. (9.5 L) Bucket | |
Handle | |
Wide-Lipped Pour Spout |
Molded | |
2.5 gal. | |
Polyethylene Coated | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1416049 | Certificate of Compliance | 24/01/2025 | 7002-0025 |
1415953 | Certificate of Compliance | 21/10/2024 | 7002-0025 |
1410452 | Certificate of Compliance | 07/08/2024 | 7002-0025 |
1404970 | Certificate of Compliance | 06/07/2024 | 7002-0025 |
1375636 | Certificate of Compliance | 28/01/2023 | 7002-0025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.