Learn More
Thermo Scientific™ Nalgene™ HDPE Heavy-Duty Bottles with Closure: Lab Pack
GREENER_CHOICE
Store and ship liquids or dry chemicals with these large, thick-walled bottles with wide mouths specially designed for extra-rugged use.
48.30 € - 81.56 €
Specifications
Color Natural
Description Heavy-Duty Bottles
Material HDPE
Closure Color White
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
View More Specs
Products 3
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10781421
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
2125-1000
1 L 53B
81.56 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
10791421
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
2125-2000
2 L 53B
48.30 €
Pack of 2
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
10606181
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
2125-4000
4 L 83B
51.05 €
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
Description
Description
Store and ship liquids or dry chemicals with these large, thick-walled and wide-mouthed Thermo Scientific™ Nalgene™ HDPE Heavy-Duty Bottles with Closure specially designed for extra-rugged use. Used as reservoirs and waste traps in automated instrumentation.
Features:
  • Leakproof† when used with Nalgene closures
  • White polypropylene linerless closures included
Specifications
Specifications
Natural
HDPE
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Wide
Heavy-duty Bottle
Polypropylene
No
Heavy-Duty Bottles
White
Yes
Round
Unlined
Screw Closure (Assembled)
Videos
SDS
Documents
Documents
Product Certifications
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ HDPE Heavy-Duty Bottles with Closure: Lab Pack