missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE, Jerry Can with Tethered Polypropylene Closure (Single Port)

Product Code. 11777825
Click to view available options
:
With single port; 3.5 gal. (13L)
This item is not returnable. View return policy

Product Code. 11777825

Brand: Thermo Scientific™ 22430013

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Ideal as a reservoir in automated systems; Closure Material: Polypropylene; Color: White

Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure, featuring a space-saving design, molded-in handle and recessed grip at the bottom to provide better control when pouring. The double-ported 2243-9013 is ideal as a reservoir in automated systems. The 53B closure can be plumbed with input/output tubing while the 38mm closure is used for refilling or emptying the reservoir.

Key Features:
  • Jerrican is ideal as a reservoir in automated systems like the PRISM™ 3700 DAN Analyzer
  • Molded-in graduations in 1 gal. and 5L increments allow for ease in dispensing
  • Molded-in handle provides a sturdy grip when carrying heavy contents
  • Integral shoulder handle and molded bottom grip make lifting comfortable
  • Wide stance and low center of gravity add stability
  • Heavy rugged design with good chemical resistance allows for a wide variety of solutions
  • Space-saving rectangular shape permits efficient storage

Ordering Information:

  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • Custom tubing sets can be designed at the Thermo Scientific Custom Tubing Set Configurator, www.thermoscientific.com/tubingconfigurator
TRUSTED_SUSTAINABILITY

Spezifikation

Capacity (English) 3.5 gal.
Capacity (Metric) 13 L
Certifications/Compliance USP Class VI Compliant/FDA21 CFR 177.1520 Compliant
Closure Color White
Closure Material Polypropylene
Closure Size 53B
Closure Type Tethered
Color White
Diameter (English) Inner Neck 1.5 in.
Diameter (Metric) Inner Neck 38 mm
For Use With (Application) Ideal as a reservoir in automated systems
Material HDPE
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Jerry Can
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt