Learn More
Thermo Scientific™ Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Spigots
Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. Spigot accepts 5/8 inch I.D. tubing. Tanks are offered in sizes from 19 to 208L.
222.00 € - 964.00 €
Specifications
Type Tank with Spigot
Material LLDPE
View More Specs
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10305661
View Documents
Thermo Scientific™
11102-0005
19 L
222.00 €
Each
In Stock
to see stock.
Add to basket
10384541
View Documents
Thermo Scientific™
11102-0007
26.5 L
267.00 €
Each
In Stock
to see stock.
Add to basket
10654141
View Documents
Thermo Scientific™
11102-0010
37.8 L
273.00 €
Each
In Stock
to see stock.
Add to basket
10603832
View Documents
Thermo Scientific™
11102-0015
56.8 L
331.00 €
Each
In Stock
to see stock.
Add to basket
10674311
View Documents
Thermo Scientific™
11102-0030
113.5 L
381.00 €
Each
In Stock
to see stock.
Add to basket
10274111
View Documents
Thermo Scientific™
11102-0055
208 L
964.00 €
Each
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Nalgene Heavy-Duty Cylindrical LLDPE Tanks include a needle-type spigot installed for draw off. This spigot accepts 5/8 inch I.D. tubing and are available in sizes ranging from 19L to 208L.
Features:
  • Needle Type Tank Spigot (Cat. No. 6421) installed to allow draw off
  • 5/8 (15.9mm) I.D.
  • Not autoclavable
Included:
Cover
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Warranty:
90 days
Specifications
Specifications
Tank with Spigot
LLDPE
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks with Spigots