missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ HDPE, Heavy-Duty Rectangular Carboy Product Code.: 15981481
Add to basket

Thermo Scientific™ Nalgene™ HDPE, Heavy-Duty Rectangular Carboy

Product Code. 15981481
20 L, 70 mm
Click to view available options
Capacity (Metric):
20 L
Closure Size:
70 mm
This item is not returnable. View return policy

Product Code. 15981481

Brand: Thermo Scientific™ 22140050

Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Store and transport reagents.; Color: White

Store and transport reagents with the rugged Thermo Scientific™ Nalgene™ Heavy-Duty HDPE Carboy. The rectangular shape helps optimize limited storage space, and the large handle is easy to use, even with gloved hands. This heavy-duty polyethelyne carboy is both durable and space-saving.

Features:

  • Heavy walled construction for durability and chemical resistance
  • Graduated to 5 L and 1 gal. increments
  • Valve-sealing graduated/leakproof† linerless 70 mm screw closure
  • Not Autoclavable

Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Liner Unlined
Closure Size 70 mm
Closure Type Screw
Color White
Dimensions (L x W x H) 12.1 x 9.2 x 15.7 in. (323 x 234 x 399 mm)
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Material HDPE
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Rectangular Carboy
Sterility Sterile
Quantity Case of 4
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1419051 Certificate TypeCertificate of Compliance Date12/12/2024 Catalog Number2214-0050
Lot Number1298899 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2214-0050
Lot Number1402405 Certificate TypeCertificate of Compliance Date12/03/2024 Catalog Number2214-0050
Lot Number1387371 Certificate TypeCertificate of Compliance Date12/06/2023 Catalog Number2214-0050
Lot Number1374824 Certificate TypeCertificate of Compliance Date19/11/2022 Catalog Number2214-0050

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ HDPE, Heavy-Duty Rectangular Carboy >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.