Learn More
Thermo Scientific™ Nalgene™ L900 Liquid Detergent
GREENER_CHOICE
Thoroughly clean all plastic labware, with an environmentally friendly formula that will not cause crazing, stress cracking or clouding when used as directed.
Brand:  Thermo Scientific™ 900-4000
Product Code. 10769041
  • 255.00 € / Pack of 4
Explore more special offers
Description
Description
Thoroughly clean all plastic labware, especially polycarbonate, with Thermo Scientific™ Nalgene™ L900 Liquid Detergent. This environmentally friendly formula will not cause crazing, stress cracking or clouding of plastic when used as directed.
  • For hand or machine use
  • Biodegradable
  • Non-toxic
  • Low foaming
  • Specially formulated for Nalgene plastic labware
TRUSTED_SUSTAINABILITY
Specifications
Specifications
L900 Liquid Detergent; L-900 Liquid Detergent *DO NOT FREEZE* 4 Liter
4 Pk.
1.06 gal.
4 L