missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.
Brand: Thermo Scientific™ 2123-0010
139.52 EUR valid until 2025-12-31
Use promo code "24373" to get your promotional price.
This item is not returnable.
View return policy
Description
Collect, store and transport dry chemicals and environmental samples with Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure. Molded-in handgrips provide greater convenience and safety when pouring.
- Useful for collecting, storing and transporting dry chemicals, environmental samples, reagents and distilled water
- Good for freezing to -100°C (-148°F)
- Excellent chemical resistance to most acids, bases and alcohols
- Square shape saves space, handles easily
- Wide mouth gives easy access
- Polypropylene cap included
- Not liquid leakproof
- Bottle and Carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Specifications
1 gal., 4 L | |
1 gal. | |
Large Wide-Mouth Square Bottle | |
3.5 in. | |
HDPE | |
Non-sterile | |
USP Class VI; FDA - 21 CFR | |
No | |
Square | |
Wide Mouth Bottle (Large) | |
11.5 in. | |
100 mm | |
Screw Cap |
4 L | |
Translucent | |
5.6 in. | |
89 mm | |
1 | |
White | |
Yes | |
Wide | |
292 mm | |
5.66 in. | |
Polypropylene | |
142 mm | |
No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction