missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack Product Code.: 10099062
Add to basket

Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack
GREENER_CHOICE

Product Code. 10099062
40 μL
Click to view available options
Capacity (Metric):
40 μL
50 μL

Product Code. 10099062

Brand: Thermo Scientific™ 3127590001

Edge
Add to basket
Add to basket

Carry out pharmaceutical and biotech applications with these LDPE dispensing tips for dropper bottles, which feature precise drip control.

Carry out pharmaceutical and biotech applications with Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles. Tips feature precise drip control and can be used with Nalgene LDPE Dropper bottles and Colored PP Closures.

Features:
  • Low density polyethylene
  • Offered in two drop sizes (40μL or 50μL)
  • Fits Nalgene Dropper Bottles Series 312750 and 312751
  • Use with Nalgene dropper closures for color coding your reagents

Ordering Information:

Dropper Bottles Dispensing Tips are bulk packed 2000 per case. Order Nalgene dropper bottles (Cat. Nos. 312750, 312751) and colored closure (Cat. No. 312760) separately.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 40 μL
Capacity (English) 1.35 oz.
Color Clear
Material LDPE
No. per Case 2000
No. per Pack 2000
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle dispensing tips meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included Yes
Drop Size 40/50 μL
Height (Metric) 16.5 mm
Product Type Dropping Bottle Tips
For Use With (Equipment) Nalgene Controlled Dropper
Width (English) 0.39 in.
Height (English) 0.65 in.
Diameter (Metric) 11.22 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ LDPE Dispensing Tips for Dropper Bottles: Bulk Pack >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.