missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure
GREENER_CHOICE

Product Code. 11379684
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
Closure Size:
24 mm
28 mm
38-430 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

Accurately fill and pour with these narrow-mouth, LDPE low particulate/low metals bottles with closure.

Accurately pour and fill Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure, which feature a narrow mouth. These low-density polyethylene bottles include certification to contain less than 20 particles per mL at 0.3 microns or greater.

Features:

  • Certified to particulate and trace metals levels
  • Excellent chemical resistance to most acids, bases and alcohols
  • Translucent
  • Guaranteed leakproof†

Metals certified to microliter (ppb) levels <0.20Hg, <0.5 Be, <1.0 As, Cd, Pb, <2.0 Sb, Se, <5.0 Ag, Co, Cr, Cu, Mn, Tl, V, <10 Ba, Ni, Zn, <50 Mg, <75 Al, <100 Ca, Fe, K, Na. Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 16 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
Description Low Particulate/Low Metals Bottle
Diameter (Metric) Inner Neck 21 mm
Material LDPE
No. per Case 48
No. per Pack 1
Sterility Non-sterile
Closure Color Natural, translucent
Low Particulate Yes
Neck Finish 28 mm
Certifications/Compliance Certified to particulate and trace metals levels
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 170 mm
Product Type Low Particulate/Low Metals Bottle
Width (English) 2.86 in.
Height (English) 6.71 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 73 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'