missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ LDPE, Round Carboy with Spigot Product Code.: 10528081
In Stock
to see stock.
Add to basket

Thermo Scientific™ Nalgene™ LDPE, Round Carboy with Spigot
GREENER_CHOICE

Product Code. 10528081
10 L
Click to view available options
Capacity (Metric):
4 L
10 L
20 L
25 L
50 L
This item is not returnable. View return policy

Product Code. 10528081

Brand: Thermo Scientific™ 23180020

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White

Thermo Scientific™ Nalgene Round LDPE Carboys are the icon of lab water dispensing and storage. Place one at every sink and every work station to enhance your productivity. Carboys have a spigot boss molded in once piece with the carboy for convenience, strength and leak resistance. Suitable for collecting and dispensing distilled water, acids and reagents. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

  • Molded in graduations in 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles make lifting comfortable for the four largest sizes
  • Molded-in spigot boss provides strength and leak resistance
  • Leakproof†

Ordering Information:

Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.

Includes:

Polypropylene spigot and screw closure with TPE gasket † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Material Polypropylene
Closure Size 83B
Color White (Opaque)
Description Round Carboys w/Spigot
Diameter (Metric) Inner Neck 64 mm
Diameter (Metric) Outer 250 mm
For Use With (Application) Collect and dispense distilled water, acids and reagents
Graduations Molded-in graduations in 1 gal. and 5 L increments
Height (Metric) With Closure 389 mm
Includes Integral shoulder handles, Molded-in spigot boss
Material LDPE
Product Type Carboy with Spigot
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1424923 Certificate TypeCertificate of Compliance Date15/03/2025 Catalog Number2318-0020
Lot Number1288766 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2318-0020
Lot Number1310031 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2318-0020
Lot Number1414464 Certificate TypeCertificate of Compliance Date25/09/2024 Catalog Number2318-0020
Lot Number1409587 Certificate TypeCertificate of Compliance Date25/09/2024 Catalog Number2318-0020
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ LDPE, Round Carboy with Spigot >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.