missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE, Wide-Mouth Carboy with Handle

For Use With: Store and transport solids, powders and liquids; Closure Material: Polypropylene; Color: Natural

69.20 € - 109.00 €

Specifications

Autoclavable Not Autoclavable
Closure Material Polypropylene
Closure Size 100-415 mm
Color Natural
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
View More Specs

Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10261501
View accessory products View Documents
Thermo Scientific™
2234-0020
10 L
69.20 €
Each
In Stock
to see stock.
Add to basket
 
10443491
View accessory products View Documents
Thermo Scientific™
2234-0030
15 L
86.15 €
Each
In Stock
to see stock.
Add to basket
 
10667521
View Documents
Thermo Scientific™
2234-0050
20 L
109.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Store and transport solids and powders with the Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Carboys with Handles. Convenient large opening promotes ease of cleaning and dispensing and can accommodate overhead mixers while its wide-shoulder handles enable easy carrying and pouring, even with gloved hands. Carboy includes leakproof white polypropylene (PP) 100 mm screw closures.

  • Constructed of low density polyethylene
  • Graduated to contain in 1 gal. and 5 L increments for ease of use
  • Wide-mouth opening to accommodate overhead mixers and is convenient for dry materials such as solids and powders
  • Leakproof†
  • Not autoclavable
Specifications

Specifications

Not Autoclavable
100-415 mm
Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
1 Ea.
Sterile
Polypropylene
Natural
LDPE
Carboy with Handle
Videos
SDS
Documents

Documents

Product Certifications

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.