Learn More
Thermo Scientific™ Nalgene™ Metric Non-Phthalate PVC Tubing
GREENER_CHOICE
Establish a leak-tight fit with metric fittings. Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticizers. Complies with FDA CFR21 for food packaging, USP VI, Cytotox, REACH, Latex-free, RoHS, CA Prop 65
64.70 € - 259.00 €
Specifications
Certifications/Compliance Meets FDA CFR21 for food packaging, USP VI, Cytotox, REACH, Latex-free, RoHS, and CA Prop 65
Description Metric Non-Phthalate PVC Tubing
Autoclavable Autoclavable
Material PVC
Sterility Non-sterile
View More Specs
Products 12
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Overall Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Overall Length (Metric) Price Quantity & Availability  
16585140
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0406
4.0 mm 6.0 mm 25 m
64.70 €
Each
In Stock
to see stock.
Add to basket
16595140
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0407
4.0 mm 7.0 mm 25 m
80.80 €
Each
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
16505150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0508
5.0 mm 8.0 mm 25 m
112.00 €
Each
In Stock
to see stock.
Add to basket
16515150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0609
6.0 mm 9.0 mm 25 m
150.00 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
16525150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0610
6.0 mm 10.0 mm 25 m
167.00 €
Each
In Stock
to see stock.
Add to basket
16535150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0710
7.0 mm 10.0 mm 25 m
217.00 €
Each
In Stock
to see stock.
Add to basket
16545150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0811
8.0 mm 11.0 mm 25 m
184.00 €
Each
In Stock
to see stock.
Add to basket
16555150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0812
8.0 mm 12.0 mm 25 m
187.00 €
Each
In Stock
to see stock.
Add to basket
16565150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-0913
9.0 mm 13.0 mm 25 m
204.00 €
Each
In Stock
to see stock.
Add to basket
16575150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-1014
10.0 mm 14.0 mm 25 m
235.00 €
Each
In Stock
to see stock.
Add to basket
16585150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-1216
12.0 mm 16.0 mm 25 m
259.00 €
Each
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
16595150
GREENER_CHOICE
missing translation for 'viewDocuments'
Thermo Scientific™
8703-1520
15.0 mm 20.0 mm 10 m
118.00 €
Each
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Nalgene™ Metric Non-Phthalate Clear Plastic PVC Tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene 180 tubing but is free from orthophthalate DEHP plasticizers. Complies with FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and complies with California proposition 65. Resists hardening and discoloration from a wide range of laboratory chemicals and does not impart odor or taste making this tubing ideal for a wide range of non-medial, general lab, research and pharmaceutical applications.
  • Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins and plasticizers
  • Free from ortho phthalate plasticizers
  • Offered in true metric sizes to fit your lab instrumentation and equipment
  • Resists hardening and discoloration from a wide range of chemicals - demonstrates dimensional stability
  • Equivalent to Tygon™ E-3603 tubing
  • Marked with I.D. and regulatory compliance as well as at 10cm. intervals for easy identification and measurement
  • Durometer hardness is Shore A, 60
  • Does not impart odor or taste
  • Can write on with Nalgene Lab marker
  • Slips easily over glass and plastic fittings; grips securely
  • Autoclavable/Transparent
Please recycle the corrugate cartons in which your Nalgene  products are packaged.
Compliance
FDA CFR21 for food packaging, USP VI, Cytotoxicity, REACH, Latex-free, RoHS, and California Proposition 65
Specifications
Specifications
Meets FDA CFR21 for food packaging, USP VI, Cytotox, REACH, Latex-free, RoHS, and CA Prop 65
Autoclavable
Non-sterile
Clear
4 x OD bend radius
160°F
Flexible Tubing
Metric Non-Phthalate PVC Tubing
PVC
Autoclave, Gas, Chemical
60 (Shore A)
Very low
71°C