Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure: Bulk Pack
Ship and store liquid reagents using these high density, durable polyethylene bottles with closure. Recommended for customers who design, assemble and certify their own combination packaging.
339.00 € - 1382.00 €
Specifications
Color | Translucent |
---|---|
Description | IP2 Narrow-Mouth Bottles |
For Use With (Application) | Storage and Shipment of Liquid Reagents |
Material | HDPE |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. |
Product Code | Brand | Capacity (Metric) | dimClosureSize | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | dimClosureSize | Price | Quantity & Availability | ||||
10252673
|
Thermo Scientific™
312099-0001 |
30 mL |
1382.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
11329794
|
Thermo Scientific™
312099-0004 |
125 mL |
923.00 €
Pack of 500 |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
10141143
|
Thermo Scientific™
312099-0008 |
250 mL |
663.00 €
Pack of 250 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
|||||
10522045
|
Thermo Scientific™
312099-0016 |
500 mL |
468.00 €
Pack of 125 |
Estimated Shipment: 12-03-2025
Log in to see stock. |
|||||
10272673
|
Thermo Scientific™
312099-0032 |
1000 mL |
339.00 €
Pack of 50 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
|||||
Description
Ship and store liquid reagents using durable Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE IP2 Bottles with Closure. These high density polyethylene bottles are a safe, economical solution for customers who design, assemble and certify their own combination packaging.
Features:- HDPE bottle with PP closure, bulk pack
- Narrow mouth
- Complies with IATA and ICAO regulations
Ordering Information:
Bottles and closures are bulk packed in separate polybags within the same carton
Recommended for:
Storage and shipment of liquid reagents
Specifications
Translucent | |
Storage and Shipment of Liquid Reagents | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. | |
Narrow | |
IP2 Bottle |
IP2 Narrow-Mouth Bottles | |
HDPE | |
Yes | |
Round | |
Polypropylene |