missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
GREENER_CHOICE

Product Code. 10162491
Click to view available options
Product Type:
Replacement Cap
Screw Cap
Spout Cap
Stem and Draw Tube
Closure Size:
20 mm
24 mm
28 mm
38 mm
43 mm
53 mm
63 mm
Color:
Amber
Natural
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.

Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.

  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
TRUSTED_SUSTAINABILITY

Specifications

Color Amber
Description Bottle Replacement Closures
Material Polypropylene
No. per Case 12
Closure Included Yes
Mouth Wide
Shape Round
Product Type Replacement Cap
For Use With (Equipment) Nalgene narrow-mouth and wide-mouth bottles
Closure Material Polypropylene
Closure Size 53 mm
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'