missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles without Closure: Bulk Pack
GREENER_CHOICE

Product Code. 10620334
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
Neck Finish:
13-415 mm
20-415 mm
Unit Size:
Pack of 2000
This item is not returnable. View return policy

Product Code. 10620334

Brand: Thermo Scientific™ 3620029050

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Store and ship small volume, valuable reagents with natural, translucent HDPE diagnostic bottles, which are packed without closures.

Store and ship small volume, valuable reagents with Thermo Scientific™ Nalgene™ Natural HDPE Diagnostic Bottles without Closure, for use in test kits and small-volume applications. These high-density polyethylene bottles are packed without closures so they can be matched with various colored closures (Cat. No. 362150). With uniform walls resistant to splitting or puncturing, these high-density polyethylene bottles are leakproof† when assembled with Nalgene closures (ordered separately).

Features:
  • Nonsterile
  • Natural, translucent high-density polyethylene
  • Durable, uniform walls resistant to splitting or puncturing

Ordering Information:

Bottles are bulk packed without closures. Packed 2000 per case. Order colored closures separately (Cat. No. 362150).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.5 oz., 15 mL
Capacity (Metric) 15 mL
Capacity (English) 0.5 oz.
Color Natural
Description 15mL, 20-415 neck finish, bulk pack
Diameter (Metric) Inner Neck 13 mm
Material HDPE
No. per Case 2000
No. per Pack 2000
Sterility Non-sterile
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Included No
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 58 mm
Product Type Narrow Mouth Bottle
Width (English) 0.98 in.
Closure Size 20-415 mm
Diameter (Metric) 25 mm
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.