missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
GREENER_CHOICE

Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.

90.17 € - 221.00 €

Specifications

Closure Material ETFE
Material FEP
No. per Case 10
No. per Pack 2
Sterility Non-sterile
View More Specs

Alert:

Note: Must be filled to 100% of total capacity for centrifugation


Products 3
Product Code Brand Capacity (Metric) Diameter (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Diameter (Metric) Price Quantity & Availability  
10539301
GREENER_CHOICE
View Documents
Thermo Scientific™
3114-0010
10 mL 16 mm
90.17 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
 
10018700
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
3114-0030
30 mL 25.7 mm
132.00 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
 
10384831
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
3114-0050
50 mL 28.8 mm
221.00 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™, specifically suited for chloroform and phenol extractions.

Features:
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† closures prevent material loss under ordinary use for most spins <10,000 × g
  • When equipped with a Nalgene sealing cap assemblies (sold separately), 30mL and 50mL tubes can be spun up to 50,000 x g
  • Must be filled to at least 100% total capacity for proper performance. 
  • Autoclavable/Biohazard/Leakproof† when used with supplied Nalgene closure /PTFE FEP

Ordering Information:

Optional sealing cap assemblies available — DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.

Includes:

Screw caps. Additional sealing cap assembly available separately (DS3131-series).

Specifications

Specifications

ETFE
10
Non-sterile
Round
Screw Cap
No
FEP
2
Autoclavable
Round
No
Centrifuge Tube
Videos
SDS
Documents

Documents

Product Certifications

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.