Learn More
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes

View contents of these contact-clear, high-speed PPCO centrifuge tubes easily. The tubes also feature excellent mechanical strength.
Brand: Thermo Scientific™ 3119-0050
106.50 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
111.51 EUR valid until 2025-12-31
Use promo code "24083" to get your promotional price.
Alert:
Must be filled to 80% of total capacity when centrifuging
Description
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes are chemically compatible with a wider variety of lab reagents than polycarbonate centrifuge tubes and are repeately autoclavable for extended performance.
Features:
- Greater chemical resistance than polycarbonate
- Can be autoclaved repeatedly—last longer than polycarbonate tubes under conditions of repeated autoclaving
- Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges; 3119-0050 can be used up to 100,605 x g if equipped with a sealing cap
- Come with polypropylene linerless closures—easy to clean and provides leakproof protection under normal conditions up to 10,000 x g
- For leakproof assurance at speeds >10,000 x g use with sealing cap assembly (sold separately)
- Leakproof†construction ensures more secure storage of samples
Ordering Information:
Optional sealing-cap assemblies available for selected sizes. Use DS3132-0020 (for 26 and 30mL tube) and DS3132-0024 (for 50mL Tube). Actual brim capacity of 3119-0050 is 42mL.

Specifications
Polypropylene | |
Polypropylene Copolymer | |
10 | |
Autoclavable | |
105.9 mm | |
Screw Cap | |
28.8 mm | |
Centrifuge Tube |
50 mL | |
100 | |
Non-sterile | |
Round | |
Round | |
No | |
No | |
100,605 x g (with sealing cap) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1431742 | Certificate of Compliance | 26/06/2025 | 3119-0050 |
1430541 | Certificate of Compliance | 20/06/2025 | 3119-0050 |
1430049 | Certificate of Compliance | 31/05/2025 | 3119-0050 |
1426231 | Certificate of Compliance | 30/05/2025 | 3119-0050 |
1426236 | Certificate of Compliance | 27/03/2025 | 3119-0050 |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24157Expires: 2025-08-31
Days
31Hours
17Minutes
21Seconds
58 -
Thermo Scientific Nalgene
Labware Legends Promotion
Save up to 60% on Thermo Scientific™ Nalgene™ labware - durable, reusable, essential.Promo Code: 24083Expires: 2025-09-30
Days
61Hours
17Minutes
21Seconds
58New Offer Save Up to 60%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
†The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.