missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile
GREENER_CHOICE

Conduct sterile sampling, storage and shipment of reagents with these clear PETG bottles that are leakproof when used with provided Nalgene lined white HDPE closures.

260.00 € - 362.00 €

Specifications

Diameter (Metric) Inner Neck 11 mm
Material PETG
No. per Case 100
No. per Pack 20
Sterility Sterile
View More Specs

Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10672083
GREENER_CHOICE
View Documents
Thermo Scientific™
2035-0005
5 mL
270.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
10704043
GREENER_CHOICE
View Documents
Thermo Scientific™
2035-0010
10 mL
260.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
10682083
GREENER_CHOICE
View Documents
Thermo Scientific™
2035-0020
20 mL
362.00 €
Pack of 100
In Stock
to see stock.
Add to basket
 
Description

Description

Conduct sterile sampling, storage and shipment of reagents and buffer solutions using Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure. These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.

Features:
  • Sterile to 10-6 SAL
  • Non-pyrogenic and non-cytotoxic
  • 20-415 lined white high-density polyethylene closure attached to PETG vial
  • Leakproof† when used with Nalgene closures

Ordering Information:

Serum Vials are packed in shrink-wrapped trays with closures attached.

Specifications

Specifications

11 mm
100
Sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Narrow
PETG
20
20-415 mm
Yes
Sample Container
Videos
SDS
Documents

Documents

Product Certifications

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.