missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ PETG Serum Vials with Continuous Thread: Nonsterile, Shrink-Wrapped Modules

Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
Unit Size:
Pack of 1260
Pack of 1656
Pack of 612
Description
Protect against leakage, breakage, and contamination using Thermo Scientific™ Nalgene Nonsterile Serum Vials. Lighter than glass. Use with lined continuous thread closures (available separately).
Features:
- PETG
- Noncytotoxic
- Nonpyrogenic
- Packaged in trayless shrink-wrap modules
- USP VI-compliant
- Nonsterile
- Leakproof† with Nalgene lined continuous-thread closures (available separately)
Ordering Information:
Serum vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 312158).
Specifications
Specifications
| Description | Serum Vials |
| Diameter (Metric) Inner | 11 mm |
| Color | Clear |
| Diameter (Metric) Outer | 30 mm |
| Material | PETG |
| Sterility | Non-sterile |
| Volume (Metric) | 20 mL |
| No. per Case | 612 |
| No. per Pack | 153 |
| Volume (English) | 2/3 oz. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction