missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
GREENER_CHOICE

Product Code. p-4526624
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
20 mL
Closure Size:
13 mm
20 mm
Unit Size:
Pack of 1260
Pack of 1932
Pack of 3451
Pack of 612
This item is not returnable. View return policy

Product Code. 10282473

Brand: Thermo Scientific™ 3420300003

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.

Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.

Features:

  • PETG
  • Noncytotoxic
  • Nonpyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL

Ordering Information:

Serum vials are packed in trayless shrink-wrapped modules. Stoppers and caps not included.

TRUSTED_SUSTAINABILITY

Specifications

Description Serum Vials
Diameter (Metric) Inner 7 mm
Color Clear
Diameter (Metric) Outer 17 mm
Material PETG
Sterility Sterile
Volume (Metric) 3 mL
No. per Case 3451
No. per Pack 493
Volume (English) 1/10 oz.
Product Line Nalgene
Volume (Metric) Total 3 mL
Closure Type Stoppers and Caps (Not Included)
Height (Metric) 37 mm
Quantity 3451 Cases, 493 Pk.
Certifications/Compliance ISO 13485:2016
Type Serum Vial
Closure Size 13 mm
For Use With (Equipment) Stopper, Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.