missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ PETG Square Media Bottles without Closure: Sterile, Shrink-Wrapped Trays
GREENER_CHOICE

Pack and ship liquid medias, buffers and sera using these heavy-walled, durable PETG square media bottles, designed for use in filling lines.

270.68 € - 649.43 €

Specifications

Color Clear
Description Media bottles without closure
For Use With (Application) Filling Lines
Material PETG
Sterility Sterile
View More Specs

Products 6
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10212673
GREENER_CHOICE
View Documents
Thermo Scientific™
342024-0030
30 mL 20-415 mm
649.43 €
Pack of 280
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
 
10447043
GREENER_CHOICE
View Documents
Thermo Scientific™
342024-0060
60 mL 24-415 mm
468.64 €
Pack of 200
Estimated Shipment: 11-03-2025
to see stock.
Add to basket
 
10394093
GREENER_CHOICE
View Documents
Thermo Scientific™
342024-0125
125 mL 38-430 mm
331.28 €
Pack of 96
In Stock
to see stock.
Add to basket
 
10029112
GREENER_CHOICE
View Documents
Thermo Scientific™
342024-0250
250 mL 38-430 mm
292.90 €
Pack of 60
In Stock
to see stock.
Add to basket
 
10562225
GREENER_CHOICE
View Documents
Thermo Scientific™
342024-0500
500 mL 38-430 mm
270.68 €
Pack of 40
In Stock
to see stock.
Add to basket
 
10707273
GREENER_CHOICE
View Documents
Thermo Scientific™
342024-1000
1 L 38-430 mm
272.70 €
Pack of 24
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
 
Description

Description

Pack and ship liquid medias, buffers and sera using Thermo Scientific™ Nalgene PETG Square Media Bottles. Heavy-walled, durable bottles are designed for use in filling lines; closures sold separately.

Features:
  • Heavy-walled and durable
  • Shatter-resistant
  • Excellent gas barrier properties
  • Sterile 10-6 SAL
  • Molded in graduations
  • Closure supplied separately
Specifications

Specifications

Clear
Filling Lines
Sterile
Yes
Square
Media Bottle
Media bottles without closure
PETG
No
Narrow
Sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.