Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Biotainer™ Bottles and Carboys
GREENER_CHOICE
Thermo Scientific™ Nalgene™ Biotainer™ Bottles and Carboys are sterile, ready-to-use and designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
230.00 € - 8395.00 €
Specifications
Material Polycarbonate
Sterility Sterile
Temperature Range (Metric) -100°C to 100°C
Liner Material Silicone
Certifications/Compliance USP Class VI, non-cytotoxic, USP <661>, non-pyrogenic
View More Specs
Products 14
Product Code Brand Capacity (Metric) Shape Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Shape Closure Size Price Quantity & Availability  
10778658
GREENER_CHOICE
View Documents
Thermo Scientific™
3500-05
5 mL Round 20 mm
5865.00 €
Pack of 500
In Stock
to see stock.
Add to basket
16449469
GREENER_CHOICE
View Documents
Thermo Scientific™
3500-05
5 mL Round 20 mm
8395.00 €
Pack of 500
In Stock
to see stock.
Add to basket
12438887
View Documents
Thermo Scientific™
3500-20
20 mL Round 20 mm
2310.00 €
Pack of 500
In Stock
to see stock.
Edge
Add to basket
10592498
View Documents
Thermo Scientific™
3030-42
125 mL Square 38 mm
790.00 €
Pack of 50
In Stock
to see stock.
Edge
Add to basket
10084597
View Documents
Thermo Scientific™
3120-42
1 L Square 48 mm
990.00 €
Pack of 35
In Stock
to see stock.
Add to basket
16582380
GREENER_CHOICE
View Documents
Thermo Scientific™
3120-42
1 L Square 48 mm
985.00 €
Pack of 35
In Stock
to see stock.
Add to basket
10432478
View Documents
Thermo Scientific™
3233-42
2 L Square 48 mm
877.00 €
Pack of 20
In Stock
to see stock.
Edge
Add to basket
11724733
View Documents
Thermo Scientific™
3405-42
5 L Square 48 mm
480.00 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
15941621
View Documents
Thermo Scientific™
3405-16
5 L Square with Handle 48 mm
476.00 €
Pack of 6
In Stock
to see stock.
Add to basket
16416519
GREENER_CHOICE
View Documents
Thermo Scientific™
3405-16
5 L Square with Handle 48 mm
591.00 €
Pack of 6
In Stock
to see stock.
Add to basket
10309347
View Documents
Thermo Scientific™
3405-16
5 L Square with Handle 48 mm
475.00 €
Pack of 6
In Stock
to see stock.
Add to basket
11859570
View Documents
Thermo Scientific™
3410-42
10 L Square 48 mm
230.00 €
Pack of 2
In Stock
to see stock.
Edge
Add to basket
15951621
View Documents
Thermo Scientific™
3410-08
10 L Square with Handle 48 mm
295.00 €
Pack of 2
In Stock
to see stock.
Add to basket
10770160
View Documents
Thermo Scientific™
3423-42
20 L Square 48 mm
549.00 €
Pack of 3
In Stock
to see stock.
Add to basket
Description
Description
Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations. Use sterile, ready-to-use Thermo Scientific™ Nalgene™ Biotainer™ Bottles and Carboys, designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
  • Durable, translucent PC construction ensures the integrity of your sample
  • Closures feature a silicone liner for leakproof† assurance
  • Customization options are available for fluid transfer applications
  • Extensive validation documentation support
  • Suitable for use in storage conditions of -100° to +100°C
  • Not for use on hot plates
Warranty: 90 days
Specifications
Specifications
Polycarbonate
-100°C to 100°C
USP Class VI, non-cytotoxic, USP <661>, non-pyrogenic
Not Autoclavable
Narrow
Polypropylene
Sterile
Silicone
Yes
Yes
Media Storage Bottle
Videos
SDS
Documents
Documents
Product Certifications
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.