missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles with Sealing Closure

Catalog Number 10758561
134.33 EUR valid until 2024-12-31
Use promo code "23291" to get your promotional price.


Click to view available options
Nominal Capacity:
250 mL
450 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.

Ideal for large-volume processes such as cell harvesting, pelleting, and protein purification, Thermo Scientific™ Nalgene™ PC Centrifuge Bottles are molded of crystal-clear polycarbonate, which allows easy sample visualization and results in excellent mechanical strength. These centrifuge bottles are the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors.

  • Molded of polycarbonate for easy visualization of samples, these bottles are ideal for low- to moderate-speed centrifugation of biological and chemical samples
  • Polypropylene sealing cap closure with silicone gasket adds leakproof† assurance at maximum speeds or when spinning hazardous materials
  • For proper performance, bottles must be filled to 80% of total capacity

Ordering Information:
When spinning 250mL bottle in a Thermo Scientific™ Sorvall™ GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required.

TRUSTED_SUSTAINABILITY
Capacity (Metric) 450 mL
Capacity (English) 16.9 oz.
Color Clear
Material Polycarbonate
No. per Case 24
No. per Pack 4
Nominal Capacity 450 mL
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Max. Force 13,700 x g
Shape Round
Height (Metric) 159.5 mm
Product Type Centrifuge Bottle
Width (English) 2.735 in.
Length (English) 2.735 in.
Height (English) 6.29 in.
Closure Material Polypropylene, red rubber
Diameter (Metric) 69.5 mm
Closure Type Sealing Cap w/Silicone Gasket
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'