Learn More
Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks with Closure
GREENER_CHOICE
Prepare and store culture media using these shatter-proof flasks -- suitable for culturing techniques. Safer than glass flasks in shakers.
74.55 € - 113.00 €
Specifications
Material Polycarbonate
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
View More Specs
Products 3
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10237850
GREENER_CHOICE
View Documents
Thermo Scientific™
4108-0125
125 mL 33 mm
83.80 €
Pack of 6
In Stock
to see stock.
Edge
Add to basket
10587341
GREENER_CHOICE
View Documents
Thermo Scientific™
4108-0250
250 mL 38 mm
74.55 €
Pack of 4
In Stock
to see stock.
Edge
Add to basket
10100211
GREENER_CHOICE
View Documents
Thermo Scientific™
4108-0500
500 mL 43 mm
113.00 €
Pack of 4
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
Description
Description
Prepare and store culture media using shatter-proof Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks with Closure-- suitable for many culturing techniques. Safer than glass flasks in shakers. Linerless closure protects contents from spills and contamination. Rugged construction combined with light weight make these flasks a valuble element of lab safety programs.
  • Molded-in graduations make liquid levels easy to estimate
  • Noncontaminating, tight-sealing, linerless polypropylene screw closures protect contents
  • Autoclavable/Transparent/Graduated
Note: Repeated autoclaving may result in some loss of mechanical strength.  Wash and rinse well before autoclaving. 
.
Specifications
Specifications
Polycarbonate
Reusable
Narrow
Plain
Autoclavable
Yes
Non-sterile
Screw