missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Brand: Thermo Scientific™ 4250-0055
31.74 EUR valid until 2024-11-29
Use promo code "21718" to get your promotional price.
This item is not returnable.
View return policy
Description
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
- Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
- Autoclavable
Specifications
54 mm | |
7 mm | |
Autoclavable | |
61 mm |
41 mL | |
Polypropylene | |
Funnel Analytical PP 55mm | |
12/Pk.; 36/Cs. |