missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
Stopper No. 10
Stopper No. 3
Stopper No. 6
Stopper No. 7
Stopper No. 8
Unit Size:
Pack of 12
Pack of 2
Pack of 4
Pack of 6
Description
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
- Translucent wall makes liquid level easily visible
- 2000mL size has molded-in graduations
- Mouth sized to accept standard sized stoppers (stopper sold separately)
- Repeatedly autoclavable for long life and rigorous use
Specifications
Specifications
| Capacity (English) | 16 oz. |
| Description | Flask Erlenmeyer PP 500mL |
| Capacity (Metric) | 500 mL |
| Material | PPCO |
| No. per Case | 12 |
| No. per Pack | 4 |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | No |
| Mouth Style | Narrow |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction