Learn More
Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
49.90 € - 109.00 €
Specifications
Material | PPCO |
---|---|
Autoclavable | Autoclavable |
Disposable | Reusable |
Mouth Style | Narrow |
Sterility | Non-sterile |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10470001
|
Thermo Scientific™
4102-0125 |
125 mL | Stopper No. 3 |
109.00 €
Pack of 12 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
10444881
|
Thermo Scientific™
4102-0250 |
250 mL | Stopper No. 6 |
72.80 €
Pack of 6 |
In Stock
Log in to see stock. |
||||
10633151
|
Thermo Scientific™
4102-0500 |
500 mL | Stopper No. 7 | N/A | |||||
10749681
|
Thermo Scientific™
4102-1000 |
1000 mL | Stopper No. 8 |
49.90 €
Pack of 2 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
10065200
|
Thermo Scientific™
4102-2000 |
2000 mL | Stopper No. 10 |
66.25 €
Pack of 2 |
In Stock
Log in to see stock. |
||||
Description
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
- Translucent wall makes liquid level easily visible
- 2000mL size has molded-in graduations
- Mouth sized to accept standard sized stoppers (stopper sold separately)
- Repeatedly autoclavable for long life and rigorous use
Recycling Information:
Specifications
PPCO | |
Reusable | |
Non-sterile |
Autoclavable | |
Narrow | |
Plain |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.