Learn More
Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp.
Brand:  Thermo Scientific™ 6165-0002
Product Code. 11394535
  • 137.00 € / Pack of 72
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
From completely open to restricted flow to completely closed—experience precise, one-hand regulation of fluid flow with the Thermo Scientific™ Nalgene™ Polypropylene Pinch Clamp. This plastic clamp will not rust, corrode or deform. Molded in one piece with no sharp edges, it accommodates a wide range of tubing.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Case of 72
Polypropylene
Pinch clamp; Fits tubing with O.D.: 1/4 to 7/16 in.
Pinch Clamp
Fits tubing with O.D.: 1/4 to 7/16 in.