missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
GREENER_CHOICE

Product Code. 10507122
Click to view available options
Dimensions (L x W x H):
18 x 6 x 2.6 in.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10507122

Brand: Thermo Scientific™ 69100618

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Reduce breakage and chipping of glass pipette tips with the stackable and lightweight Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray.

Reduce breakage and chipping of glass pipette tips with the Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray. Horizontal design allows pipettes to be fully immersed in disinfectant. Rigid plastic tray is stackable, lightweight, quiet and less expensive than stainless steel. Cover fits into groove of pan to minimize spillage of hazardous materials.

  • Holds pipets up to 40cm long in a stackable, lightweight alternative to stainless steel
  • Can also be used to sterilize small labware and instruments—resists aggressive disinfectants
  • Has groove for cover to minimize spillage of hazardous materials and a hand grip on the cover
  • Autoclavable

Compliance:

  • Meets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens.
  • Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of pipets are required before cleaning and disposal

Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Meets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens. Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of
Dimensions (L x W x H) 18 x 6 x 2.6 in.
Description Sterilizing tray with rigid plastic construction
Height (Metric) 6.7 cm
Length (Metric) 46 cm
Material Polypropylene
Type Sterilizing Tray
Width (Metric) 15 cm
For Use With Holds pipets up to 40cm long
Length (English) 18 in.
No. per Pack 1
Height (English) 2.6 in.
No. per Case 6
Shape Rectangular
Width (English) 6 in.
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.