missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Polypropylene Pour Spout
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Click to view available options
Description:
Pour Spout
For Use With (Equipment):
Nalgene bottles with 38-430mm neck finish
Description
Enjoy easy one-handed liquid dispensing without leaving bottles completely open to the environment. Thermo Scientific™ Nalgene™ Polypropylene Pour Spout is made to fit all Nalgene bottles with 38-430 neck finish. Vented to allow smooth flow for easy liquid dispensing.
- Used for all Nalgene bottles with 38-430 neck finish
- Allows easy one-handed liquid dispensing without leaving bottles completely open to the environment
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Color | Translucent |
Description | Pour Spout |
Material | Polypropylene |
No. per Case | 6 |
No. per Pack | 6 |
Liner Material | Polyethylene |
Product Type | Pouring Spout |
For Use With (Equipment) | Nalgene bottles with 38-430mm neck finish |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction