Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Rectangular Carboy with Closure
Description
Store solutions and handle large volumes of powders and other solid samples with Thermo Scientific™ Nalgene™ Rectangular Polypropylene Carboys with Closure. Featuring a space-saving design, a wide mouth opening for filling and cleaning and a sturdy stainless-steel handle, these carboys are tough, translucent and autoclavable.
Features:
- Wide-mouth opening for easy filling, transferring and cleaning of carboys
- Molded in graduations in 1 gallon and 5 L increments for ease in dispensing
- Stainless-steel handle for ease in transporting containers
- Autoclavable, graduated and leakproof†
Includes:
- Polypropylene screw closure

Specifications
Specifications
Autoclavable | Autoclavable |
Capacity (English) | 2.4 gal. |
Capacity (Metric) | 9 L |
Closure Material | Polypropylene |
Closure Size | 100-415 mm |
Color | White |
Description | Autoclavable Rectangular Carboys |
Dimensions (L x W x H) | 9 x 6 x 14 in. (220 x 153 x 360 mm) |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Material | Polypropylene |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1423782 | Certificate of Compliance | 06/03/2025 | 2212-0020 |
1288319 | Certificate of Compliance | 10/12/2024 | 2212-0020 |
1285186 | Certificate of Compliance | 10/12/2024 | 2212-0020 |
1305778 | Certificate of Compliance | 10/12/2024 | 2212-0020 |
1416956 | Certificate of Compliance | 07/11/2024 | 2212-0020 |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.