missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels with Closure
GREENER_CHOICE

Product Code. p-4525026
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 4
This item is not returnable. View return policy

Product Code. 10238970

Brand: Thermo Scientific™ 43000125

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Perform a variety of lab duties with chemically inert, translucent and autoclavable polypropylene separatory funnels.

Perform a variety of liquid phase separations with translucent and autoclavable Thermo Scientific™ Nalgene™ Polypropylene Copolymer (PPCO) Separatory Funnels. Good chemical compatibility with a wide range of laboratory solutions involved in liquid-liquid extractions. Polypropylene closure and TFE stopcock require no lubrication. These separatory funnels are impact resistant and won't chip or shatter, making them a valuable asset to lab safety programs.

Features:

  • Separation lines between immiscible liquids are easily observed
  • Phase interface is observable down to the stopcock
  • TFE stopcocks need no lubrication, allowing for easier handling and operation
  • Funnel is chemically compatible with a wide range of lab reagents
  • Autoclavable/Leakproof

Includes:

  • Polypropylene funnel, Teflon™ TFE stopcock and housing, polypropylene screw closure
Ordering Information:
  • For replacement stopcock, order Cat. No. 714301-1025
Notes:
  • Funnels are susceptible to softening and swelling from extended exposure to aggressive solvents though the effect is reversible.
  • Stem lengths, 65mm. Autoclavable except for stopcock assembly, which is easily removed for chemical sterilization.
  • Autoclavable except for stopcock assembly, which is easily removed and chemically sterilized.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Material Polypropylene Copolymer (PPCO)
Height (Metric) 258 mm
Description 26mm
Quantity 1/Pk.; 6/Cs.
Product Type Separatory Funnel

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.