missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure Product Code.: 10373401
Add to basket

Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure
GREENER_CHOICE

Product Code. 10373401
2 L, 63 mm
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2 L
Closure Size:
28 mm
38 mm
48 mm
53 mm
63 mm
This item is not returnable. View return policy

Product Code. 10373401

Brand: Thermo Scientific™ 20070064

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Transport, sample or store liquids and dry materials with these space-efficient bottles featuring excellent chemical resistance to most acids, bases and alcohols.

Transport, sample or store liquids and dry materials with these space-efficient bottles that feature excellent chemical resistance to most acids, bases and alcohols. Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure take up less shelf space than traditional round bottles. HDPE bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Rectangular shape makes it easy to fit in freezer for storage up to -100°C (-148°F)
  • Linerless polypropylene screw closures provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Leakproof†

Includes:

Polypropylene screw caps

Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 64 oz., 2000 mL
Capacity (Metric) 2 L
Capacity (English) 64 oz.
Color Translucent
Description Wide-Mouth Rectangular Bottle
Diameter (English) Inner Neck 2 in.
Diameter (Metric) Inner Neck 51 mm
Material HDPE
Length (Metric) 152 mm
No. per Case 12
No. per Pack 4
Sterility Non-sterile
Width (Metric) 84 mm
Closure Color Natural, translucent
Certifications/Compliance USP Class VI; FDA - 21 CFR
Closure Included Yes
Graduated No
Mouth Wide
Shape Rectangular
ACT Label Yes
Height (Metric) 242 mm
Product Type Wide Mouth Bottle
Height (English) 9.5 in.
Closure Liner Unlined
Closure Material Polypropylene
Closure Size 63 mm
Diameter (Metric) 152.4 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.