missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot Product Code.: 10008080
Add to basket

Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot
GREENER_CHOICE

Product Code. 10008080
20 L
Click to view available options
Capacity (Metric):
9 L
20 L
This item is not returnable. View return policy

Product Code. 10008080

Brand: Thermo Scientific™ 23220050

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Ideal for use when visual inspection of contents is necessary; Autoclavable; Closure Material: Polypropylene; Color: Clear

Easily fill and clean the Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ with Spigot, which features a wide-mouth opening, a sturdy stainless-steel handle and space-saving rectangular design. Clear construction allows easy viewing of carboy contents. Super tough; impact resistant. Ideal for refrigerated or frozen storage of aqueous solutions, or for dispensing sterile water, aqueous reagents and media. Nalgene polycarbonate carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features

  • Molded-in graduations in 1 gallon and 5L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Molded-in spigot boss provides strength and leak resistance
  • A clear wall makes this carboy ideal when visual inspection of contents is necessary
  • Follow autoclave instructions provided with the product for proper function; tubing adapter is not autoclavable
  • 20L size has molded-in hand grips in the sidewall for easier handling
  • Autoclavable/Transparent/Graduated

Includes:

Polypropylene spigot and screw closure

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Closure Size 100 mm
Color Transparent
Description Autoclavable Clear Rectangular Carboy w/Spigot
Diameter (Metric) Inner Neck 86 mm
For Use With (Application) Ideal for use when visual inspection of contents is necessary
Height (Metric) With Closure 399 mm
Includes Molded-in spigot boss w/tubing adapter
Length (Metric) 229 mm
Material Polycarbonate
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Spigot
Width (Metric) 320 mm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1301997 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2322-0050
Lot Number1411444 Certificate TypeCertificate of Compliance Date14/09/2024 Catalog Number2322-0050
Lot Number1401045 Certificate TypeCertificate of Compliance Date08/02/2024 Catalog Number2322-0050
Lot Number1389420 Certificate TypeCertificate of Compliance Date16/07/2023 Catalog Number2322-0050
Lot Number1379787 Certificate TypeCertificate of Compliance Date27/02/2023 Catalog Number2322-0050
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Rectangular Polycarbonate Clearboy™ Carboy with Spigot >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.