Learn More
Thermo Scientific™ Nalgene™ Rectangular Polypropylene Tank with Cover
Significantly reduce evaporation and contamination by using these covered, rectangular tanks, which are offered in sizes from 8 to 114L to meet application needs.
253.00 € - 687.00 €
Specifications
Type Tank with Cover
Material Polypropylene
View More Specs
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10232611
View Documents
Thermo Scientific™
14200-0002
8 L
253.00 €
Each
In Stock
to see stock.
Add to basket
10683961
View Documents
Thermo Scientific™
14200-0005
20 L
458.00 €
Each
In Stock
to see stock.
Add to basket
10544251
View Documents
Thermo Scientific™
14200-0010
30 L
307.00 €
Each
In Stock
to see stock.
Add to basket
10409382
View Documents
Thermo Scientific™
14200-0015
38 L
383.00 €
Each
In Stock
to see stock.
Add to basket
10008770
View Documents
Thermo Scientific™
14200-0020
50 L
569.00 €
Each
In Stock
to see stock.
Add to basket
10708181
View Documents
Thermo Scientific™
14200-0045
120 L
687.00 €
Each
Estimated Shipment: 19-01-2026
to see stock.
Add to basket
Description
Description
Significantly reduce evaporation and contamination by using a Thermo Scientific™ Nalgene™ Rectangular Polypropylene Tank with Cover, offered in sizes ranging from 8 to 114L. Tanks can be modified with a factory-installed spigot.
Features:
  • Suitable for use with many organic chemicals
  • Offers good stress-crack resistance
  • Autoclavable
Warranty:
90 days
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
Specifications
Specifications
Tank with Cover
Polypropylene
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Rectangular Polypropylene Tank with Cover