Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes; PP screw closure, silicone gasket
Ensure leakproof service at their maximum speeds >10,000 x g, even when full
43.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10420533
|
Thermo Scientific™
713132-0063 |
Replacement O-ring (for Mfr. No. DS3132-0063) |
43.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11969436
|
Thermo Scientific™
713132-0058 |
Replacement O-ring (for Mfr. No. DS3132-0058) |
43.00 €
Pack of 10 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
Description
- Polypropylene closures with silicone gasket
Before autoclaving, set closure on top of the container without engaging the threads.
Specifications
Autoclavable | |
Silicone | |
Silicone |