Learn More
Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Brand: Thermo Scientific™ 4116-0125
221.90 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Save preparation time and avoid contamination risk by using Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
- Sterile and single use to reduce contamination risk
- Collapses when autoclaved to reduce bio hazardous waste
- Graduated for quick volume assessment
- Excellent validation support: Validation binder available upon request†
Note:
† Validation binders are available to customers as support documentation during your product validation process. This confidential information can be requested at: rocregsupport@thermofisher.com. Following your request submission, a Confidentiality Agreement will be forwarded to you for completion.

Specifications
4 oz. | |
PETG | |
1 | |
None | |
Single-use | |
Narrow | |
Baffled | |
38-430 mm |
125 mL | |
24 | |
Erlenmeyer | |
Not Autoclavable | |
Yes | |
Sterile | |
Vented |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.