missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Spigot Closure with Polypropylene Lock Nut and TPE plug Product Code.: 12807840
Add to basket

Thermo Scientific™ Nalgene™ Polypropylene, Spigot Closure with Polypropylene Lock Nut and TPE plug
GREENER_CHOICE

Product Code. 12807840
Spigot Closure PP/TPE fits all 1-1/8 in. spigot bosses 1-1/8 inch, 16 mm
Click to view available options
Description:
Spigot Closure PP/TPE fits all 1-1/8 in. spigot bosses 1-1/8 inch
Diameter (Metric) Inner Tubing:
16 mm
This item is not returnable. View return policy

Product Code. 12807840

Brand: Thermo Scientific™ 64230010

Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: Nalgene carboys, Clearboys, Lowboys and tanks w/thread size of 1-1/8 in., 12 male screw head

Use Thermo Scientific™ Nalgene™ Spigot Closure with Polypropylene Lock Nut and TPE Plug in place of a spigot when autoclaving liquids in a Nalgene carboy to provide a leakproof† seal. This closure is also useful when handling could damage or accidentally turn on a spigot, or when a spigot-less carboy is required. Fits all Nalgene carboys with spigot and selected Nalgene tanks with spigot.

Features:
  • Closure keeps solutions safe by providing a leakproof† seal
  • Designed to fit Nalgene carboys, Clearboys™, Lowboys™ and tanks that have a thread size of 1-1/8″, 12 male screw head
  • Autoclavable

Includes:

White polypropylene lock nut, TPE plug

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

TRUSTED_SUSTAINABILITY

Specifications

Description Spigot Closure PP/TPE fits all 1-1/8 in. spigot bosses 1-1/8 inch
Diameter (Metric) Inner Tubing 16 mm
For Use With (Equipment) Nalgene carboys, Clearboys, Lowboys and tanks w/thread size of 1-1/8in., 12 male screwhead
Material Polypropylene, TPE
No. per Case 24/Cs.
No. per Pack 4 Pk.
Product Type Spigot Closure

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Polypropylene, Spigot Closure with Polypropylene Lock Nut and TPE plug >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.