missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
Description
Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Eliminates costly washing, depyrogenation and autoclaving steps
- Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
- Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
- Supported by extensive validation documentation
Specifications
Specifications
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | Clear |
Diameter (English) | 2.13 in. |
Diameter (English) Inner Neck | 1.10 in. |
Diameter (Metric) Inner Neck | 28 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product |
Material | PETG |
No. per Case | 50/Cs. |
No. per Pack | 5/Pk. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'