Learn More
Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle
Description
Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.
- Eliminates costly washing, depyrogenation and autoclaving steps
- Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
- Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
- Supported by extensive validation documentation

Specifications
Specifications
Capacity (Metric) | 2 L |
Capacity (English) | 64 oz. |
Color | Clear |
Diameter (English) | 4.6 in. |
Diameter (English) Inner Neck | 1.50 in. |
Diameter (Metric) Inner Neck | 39 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation , Validation Testing, Drug Substance, Drug Product |
Height (Metric) With Closure | 271 mm |
Material | PETG |
No. per Case | 20/Cs. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1398991 | Certificate of Compliance | 04/02/2025 | CE-N2019-2000 |
1420822 | Certificate of Compliance | 22/01/2025 | CE-N2019-2000 |
1419684 | Certificate of Compliance | 22/01/2025 | CE-N2019-2000 |
7315463010 | Certificate of Compliance | 10/12/2024 | CE-N2019-2000 |
1337056 | Certificate of Compliance | 10/12/2024 | CE-N2019-2000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.