missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Square PETG Platinum Certified Clean Media Bottle

Product Code. 16480601
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ Nalgene™ Square PETG Media Bottles are certified sterile, non-pyrogenic, and ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.

Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media. Certified and lot-to-lot tested to contain less than 1/3 the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the container/closure, including two heat-sealed sterile barriers to protect product prior to use.

  • Eliminates costly washing, depyrogenation and autoclaving steps
  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates
  • Also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
  • Supported by extensive validation documentation
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (English) 1.5 in.
Diameter (English) Inner Neck 0.55 in.
Diameter (Metric) Inner Neck 14 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Height (Metric) With Closure 64 mm
Material PETG
No. per Case 70/Cs.
No. per Pack 7/Pk.
Sterility Sterile
Temperature Range (Metric) -70°C to 70°C
Weight (English) 3.2 lb.
Low Particulate <1/3 USP <788> (Ultralow particulate)
Certifications/Compliance USP Class VI Compliant, Certified Radiation Stabilized (SAL 10-6)
Autoclavable Not Autoclavable
Mouth Narrow
Height (Metric) 64 mm
Product Type Media Bottle
Width (English) 6.9 in.
Height (English) 2.52 in.
Closure Size 20 mm
Diameter (Metric) 38 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'