Learn More
Thermo Scientific™ Nalgene™ Staining Box
GREENER_CHOICE
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods with these staining boxes.
44.90 €
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10492491
GREENER_CHOICE
View Documents
Thermo Scientific™
5705-1010
Gel Staining Box PMP 12.5 x 12.5 x 5 cm
44.90 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods. Thermo Scientific™ Nalgene™ Staining Box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 4.8 in. x 4.8 in (189 mm x 189 mm) gel along with 500mL of liquid.
  • Polymethylpentene box and polypropylene copolymer cover
  • Useful over a -70° to +70°C (-94° to +158°F) temperature range
  • Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
  • LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
  • Plug won't interfere with cover or contents of boxes on a benchtop or shaker
  • Stackable, with a handle on each end for safe, easy carrying
  • Microwavable and easy to clean
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.