Learn More
Thermo Scientific™ Nalgene™ Stopcocks
Develop in-house fluid transfer systems for accurate filling or dispensing. These stopcocks provide reliable, accurate liquid flow control.
Brand: Thermo Scientific™ 6460-0004
Description
Permit reliable, accurate liquid flow control in liquid handling systems by using Thermo Scientific™ Nalgene™ Stopcocks. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.
- Serrated tubulation on each end accepts 1/2″ through 5/16″ tubing
- Polypropylene with Teflon™ TFE plug
- Autoclavable
Specifications
Stopcock | |
Polypropylene | |
6 | |
Polypropylene |
4 mm | |
1 | |
In-House Fluid Transfer System |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7242957010-COC | Certificate of Analysis | 13/04/2017 | 6460-0004 |
7102352060-COC | Declaration of Conformity | 31/03/2017 | 6460-0004 |
7216500010-COC | Declaration of Conformity | 24/03/2017 | 6460-0004 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.