missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Storm Water Samplers and Mounting Kit
GREENER_CHOICE

A convenient and affordable device for collecting quarterly first flush storm water grab samples in compliance with EPA sampling requirements.

Brand:  Thermo Scientific™ 1100-1000

 View more versions of this product

Product Code. 11349754

  • 233.00 € / Pack of 4
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Nalgene™ Storm Water Sampler is a convenient and affordable device for collecting quarterly storm water grab samples in compliance with EPA sampling requirements. No more standing in the rain waiting for water to flow or missed sampling events. The Storm Water Sampler can collect a full one liter grab sample of first flush storm water runoff through a storm water ditch, stream or storm grate outfall. Simply position the sampler in its protective mounting tube prior to a rain event, and leave it in place until after the storm.

Features:
  • Leakproof† when polypropylene closure is applied
  • Disposable sampling device is intended for one-time use
  • Ideal for collecting samples for inorganic analyses

Includes:

EPDM gasket, polypropylene ball valve and closure

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Water Sampler
Storm Water Sampler 1L HDPE
33.8 oz.
63 mm
1/Pk., 4/Cs.
HDPE
Polypropylene
1 L
Meets EPA NPDES MSGP (Fed. Reg. FRL-6880-5 10/20/00 and most state grab sampling requirements
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers