Learn More
Product Code | Brand | Diameter (Metric)Top | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Capacity (Metric) | Price | Quantity & Availability | |||||
10365701
![]() |
Thermo Scientific™
4256-0234 |
62 mm | 62 mL |
36.55 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
10709041
![]() |
Thermo Scientific™
4256-0414 |
98 mm | 270 mL |
44.70 €
Pack of 12 |
Estimated Shipment: 20-06-2025
Log in to see stock. |
|||||
10440723
![]() |
Thermo Scientific™
4256-0638 |
150 mm | 1059 mL |
161.00 €
Pack of 12 |
In Stock
Log in to see stock. ![]() |
|||||
Description
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.
- 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
- 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and High Density Polyethylene (HDPE) labware at end of product life in communities where these materials can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Utility Funnels