Learn More
Thermo Scientific™ Nalgene™ Rubber Stopper, for Vacuum Manifold
GREENER_CHOICE
Replace the parts and accessories on the Nalgene™ Vacuum Manifold with the Nalgene Vacuum Manifold Accessories.
Brand:  Thermo Scientific™ DS0396-0080
42.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12085097
  • 44.05 € / Pack of 3
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
  • Compatible with Nalgene Vaccum Manifold (Cat. No. DS0345-0001)
  • Rubber stopper, number 8
  • 5/8 in. hole
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15.875 mm
Rubber
8
Nalgene Vacuum Manifold (Cat. No. DS0345-0001)
Rubber Stopper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Rubber Stopper, for Vacuum Manifold > Number 8