Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure: Bulk Pack
Store and ship liquid reagents with these leakproof, wide-mouth bottles with closure that meet ICAO and IATA requirements.
524.19 € - 1366.53 €
Specifications
Color | White |
---|---|
Description | IP2 Wide-Mouth Bottles |
For Use With (Application) | Storage and Shipment of Liquid Reagents |
Material | HDPE |
Certifications/Compliance | Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10190563
|
Thermo Scientific™
312199-0004 |
125 mL | 38-415 mm |
1366.53 €
Pack of 500 |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||||
10212533
|
Thermo Scientific™
312199-0008 |
250 mL | 43-415 mm |
1037.27 €
Pack of 250 |
In Stock
Log in to see stock. |
||||
10484933
|
Thermo Scientific™
312199-0016 |
500 mL | 53-415 mm |
717.10 €
Pack of 125 |
In Stock
Log in to see stock. |
||||
10521085
|
Thermo Scientific™
312199-0032 |
1000 mL | 63-415 mm |
524.19 €
Pack of 50 |
In Stock
Log in to see stock. |
||||
Description
Store and ship liquid reagents with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE IP2 Bottles with Closure. Leakproof† when used with Nalgene closures and constructed of durable high-density polyethylene, these safe, economical bottles are recommended for those designing, assembling and certifying their own combination packaging.
Features:- HDPE bottles with PP closures
- Wide-mouth
- Complies with IATA and ICAO regulations
- Leakproof† when used with Nalgene closures
Ordering Information:
Bottles and closures are bulk packed in separate polybags within the same carton
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance DatabaseSpecifications
White | |
Storage and Shipment of Liquid Reagents | |
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Bottles are evaluated at 15psi (103kPa) per 49 CFR 173.27 (c)(2), ICAO Technical Instructions Part 4; 1.1.6, and IATA Dangerous Goods Regulations Section 5.0.2.9. | |
Wide | |
IP2 Bottle |
IP2 Wide-Mouth Bottles | |
HDPE | |
Yes | |
Round | |
Polypropylene |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.